siRNA Details
| Gene Symbol: 4930430F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045186.1 | 3, 4 | 481 | |||
| GenBank mRNA | AK015252.1 | - | 3, 4 | 489 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714809.1 | - | 3, 4 | 483 | Chr. 15 - 31039136 - 31043333 [+] on Build GRCm38 | |

4930430F21Rik
Genomic Map