siRNA Details
| Gene Symbol: Gm12371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081400.1 | - | 2 | 951 | ||
| BB272510.1 | - | 1 | 188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12371| Gene Symbol: Gm12371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081400.1 | - | 2 | 951 | ||
| BB272510.1 | - | 1 | 188 | |||