siRNA Details
| Gene Symbol: Gm12371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081400.1 | - | 2 | 518 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB648387.1 | - | 2 | 518 | Chr. 4 - 36952966 - 36987589 [+] on Build GRCm38 | |

Gm12371
Genomic Map