siRNA Details
Gene Symbol: 4930444F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038034.1 | 3 | 2293 | |||
GenBank mRNA | AK029596.1 | - | 3 | 2281 | ||
EU523860.1 | - | 2 | 2045 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930444F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038034.1 | 3 | 2293 | |||
GenBank mRNA | AK029596.1 | - | 3 | 2281 | ||
EU523860.1 | - | 2 | 2045 |