siRNA Details
| Gene Symbol: 4930444F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038034.1 | 3 | 2170 | |||
| GenBank mRNA | AK029596.1 | - | 3 | 2158 | ||
| EU523860.1 | - | 2 | 1922 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930444F02Rik| Gene Symbol: 4930444F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038034.1 | 3 | 2170 | |||
| GenBank mRNA | AK029596.1 | - | 3 | 2158 | ||
| EU523860.1 | - | 2 | 1922 | |||