siRNA Details
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020006.1 | - | 1 | 1196 | ||
AK036657.1 | - | 3 | 2475 | |||
AK080208.1 | - | 1 | 1701 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020006.1 | - | 1 | 1196 | ||
AK036657.1 | - | 3 | 2475 | |||
AK080208.1 | - | 1 | 1701 |