siRNA Details
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020006.1 | - | 1 | 2145 | ||
AK036657.1 | - | 3 | 3426 | |||
AK080208.1 | - | 1 | 2650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB223075.2 | - | 1 | 551 | Chr. 11 - 98513785 - 98514446 [-] on Build GRCm38 |
