siRNA Details
| Gene Symbol: Gm33104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029599.1 | - | 1 | 47 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614960.1 | - | 1 | 47 | Chr. 10 - 19486715 - 19507243 [+] on Build GRCm38 | |

Gm33104
Genomic Map