siRNA Details
| Gene Symbol: 4931422A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016472.1 | - | 2 | 703 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716148.1 | - | 2 | 703 | Chr. 2 - 104025941 - 104028361 [-] on Build GRCm38 | |

4931422A03Rik
Genomic Map