siRNA Details
| Gene Symbol: 6330412A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018162.1 | - | 1 | 2467 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV326564.1 | - | 139 | Chr. - - [-] on Build | ||

6330412A17Rik
Genomic Map