siRNA Details
Gene Symbol: Gm30246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135432.1 | - | 2 | 558 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ051840.1 | - | 2 | 558 | Chr. 14 - 76780827 - 76781874 [+] on Build GRCm38 |
