siRNA Details
Gene Symbol: 5730480H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045500.1 | 1 | 789 | |||
GenBank mRNA | AK017706.1 | - | 1 | 789 | ||
AK141983.1 | - | 3525 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5730480H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045500.1 | 1 | 789 | |||
GenBank mRNA | AK017706.1 | - | 1 | 789 | ||
AK141983.1 | - | 3525 |