siRNA Details
| Gene Symbol: 2900001G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013461.1 | - | 1 | 1395 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF470096.1 | - | 1 | 418 | Chr. 17 - 30939847 - 30940369 [+] on Build GRCm38 | |

2900001G08Rik
Genomic Map