siRNA Details
| Gene Symbol: B230337E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046039.1 | - | 1 | 844 | ||
| AK144670.1 | - | 1 | 1254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230337E12Rik| Gene Symbol: B230337E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046039.1 | - | 1 | 844 | ||
| AK144670.1 | - | 1 | 1254 | |||