siRNA Details
| Gene Symbol: 1700028E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045699.1 | 4 | 576 | |||
| GenBank mRNA | AK042042.1 | - | 4 | 543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700028E10Rik| Gene Symbol: 1700028E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045699.1 | 4 | 576 | |||
| GenBank mRNA | AK042042.1 | - | 4 | 543 | ||