siRNA Details
| Gene Symbol: 1700028E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045699.1 | 3 | 378 | |||
| GenBank mRNA | AK042042.1 | - | 3 | 345 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB638822.1 | - | 3 | 342 | Chr. 5 - 151368711 - 151394716 [+] on Build GRCm38 | |

1700028E10Rik
Genomic Map