siRNA Details
Gene Symbol: E130215H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040331.1 | 1, 2 | 467 | |||
GenBank mRNA | AK042041.1 | - | 1, 2 | 162 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E130215H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040331.1 | 1, 2 | 467 | |||
GenBank mRNA | AK042041.1 | - | 1, 2 | 162 |