siRNA Details
| Gene Symbol: 1700016G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045891.1 | 3 | 433 | |||
| GenBank mRNA | AK006031.1 | - | 3 | 434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700016G22Rik| Gene Symbol: 1700016G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045891.1 | 3 | 433 | |||
| GenBank mRNA | AK006031.1 | - | 3 | 434 | ||