siRNA Details
| Gene Symbol: 2900075N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032471.1 | - | 1 | 1333 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ444147.1 | - | 1 | 161 | Chr. X - 101324744 - 101325451 [+] on Build GRCm38 | |

2900075N08Rik
Genomic Map