siRNA Details
| Gene Symbol: Gm15155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028243.1 | - | 2, 3 | 253 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712647.1 | - | 2, 3 | 253 | Chr. X - 155624779 - 156343166 [+] on Build GRCm38 | |

Gm15155
Genomic Map