siRNA Details
| Gene Symbol: 1700083H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006983.1 | - | 1 | 81 | ||
| AK019662.1 | - | 1 | 38 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700083H02Rik| Gene Symbol: 1700083H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006983.1 | - | 1 | 81 | ||
| AK019662.1 | - | 1 | 38 | |||