siRNA Details
| Gene Symbol: 1700083H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006983.1 | - | 2 | 290 | ||
| AK019662.1 | - | 2 | 246 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715215.1 | - | 2 | 246 | Chr. 12 - 71383454 - 71384166 [-] on Build GRCm38 | |

1700083H02Rik
Genomic Map