siRNA Details
| Gene Symbol: 5430434G16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017390.1 | - | 1 | 410 | ||
| AK139147.1 | - | 1 | 410 | |||
| BE373635.1 | - | 1 | 394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430434G16Rik| Gene Symbol: 5430434G16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017390.1 | - | 1 | 410 | ||
| AK139147.1 | - | 1 | 410 | |||
| BE373635.1 | - | 1 | 394 | |||