siRNA Details
| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035958.1 | - | 1 | 3032 | ||
| AK053482.1 | - | 1 | 1265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcor3| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035958.1 | - | 1 | 3032 | ||
| AK053482.1 | - | 1 | 1265 | |||