siRNA Details
| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035958.1 | - | 1 | 2685 | ||
| AK053482.1 | - | 1 | 918 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB539863.2 | - | 1 | 220 | Chr. 1 - 192128108 - 192128750 [-] on Build GRCm38 | |

Rcor3
Genomic Map