siRNA Details
| Gene Symbol: Sema6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078490.1 | - | 1 | 3168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB684832.1 | - | 1 | 73 | Chr. 18 - 47281687 - 47282100 [-] on Build GRCm38 | |

Sema6a
Genomic Map