siRNA Details
| Gene Symbol: C330004P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021178.1 | - | 3 | 452 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728731.1 | - | 3 | 452 | Chr. 10 - 15791777 - 15807069 [+] on Build GRCm38 | |

C330004P14Rik
Genomic Map