siRNA Details
| Gene Symbol: AI606473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040387.1 | 1 | 213 | |||
| GenBank mRNA | AK142074.1 | - | 1 | 214 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730156.1 | - | 1 | 210 | Chr. 3 - 154330824 - 154333167 [+] on Build GRCm38 | |

AI606473
Genomic Map