siRNA Details
| Gene Symbol: AI606473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040387.1 | 3 | 1991 | |||
| GenBank mRNA | AA387793.1 | - | 1 | 326 | ||
| AK142074.1 | - | 3 | 1990 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI606473| Gene Symbol: AI606473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040387.1 | 3 | 1991 | |||
| GenBank mRNA | AA387793.1 | - | 1 | 326 | ||
| AK142074.1 | - | 3 | 1990 | |||