siRNA Details
| Gene Symbol: I730030J21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045781.1 | 1 | 54 | |||
| NR_045782.1 | 1 | 54 | ||||
| GenBank mRNA | AK146183.1 | - | 1 | 55 | ||
| AK163102.1 | - | 1 | 38 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

I730030J21Rik| Gene Symbol: I730030J21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045781.1 | 1 | 54 | |||
| NR_045782.1 | 1 | 54 | ||||
| GenBank mRNA | AK146183.1 | - | 1 | 55 | ||
| AK163102.1 | - | 1 | 38 | |||