siRNA Details
| Gene Symbol: Gm40918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048601.1 | - | 128 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU946228.1 | - | 2 | 131 | Chr. 13 - 37692055 - 37717648 [-] on Build GRCm38 | |

Gm40918
Genomic Map