siRNA Details
| Gene Symbol: LOC102640923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158984.1 | - | 2, 3 | 176 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY285137.1 | - | 2, 3 | 176 | Chr. 8 - 76797053 - 76899985 [-] on Build GRCm38 | |

LOC102640923
Genomic Map