siRNA Details
| Gene Symbol: Cdyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085145.1 | - | 1 | 743 | ||
| AK172549.1 | - | 1 | 1352 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU922735.1 | - | 1 | 28 | Chr. 8 - 116572656 - 116573252 [-] on Build GRCm38 | |

Cdyl2
Genomic Map