siRNA Details
| Gene Symbol: Gm15577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035866.1 | - | 195 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB628526.1 | - | 2 | 195 | Chr. 3 - 156905413 - 156920780 [-] on Build GRCm38 | |

Gm15577
Genomic Map