siRNA Details
| Gene Symbol: 2010004M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008102.1 | - | 1 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010004M13Rik| Gene Symbol: 2010004M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008102.1 | - | 1 | 403 | ||