siRNA Details
| Gene Symbol: 4933422A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040715.1 | 5 | 1088 | |||
| GenBank mRNA | AK016867.1 | - | 5 | 1089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933422A05Rik| Gene Symbol: 4933422A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040715.1 | 5 | 1088 | |||
| GenBank mRNA | AK016867.1 | - | 5 | 1089 | ||