siRNA Details
| Gene Symbol: 4933422A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040715.1 | 5 | 863 | |||
| GenBank mRNA | AK016867.1 | - | 5 | 864 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716546.1 | - | 5 | 864 | Chr. 9 - 35281105 - 35285037 [+] on Build GRCm38 | |

4933422A05Rik
Genomic Map