siRNA Details
| Gene Symbol: 1700127F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018992.1 | - | 1 | 275 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707401.1 | - | 1 | 275 | Chr. 12 - 112875211 - 112875814 [-] on Build GRCm38 | |

1700127F24Rik
Genomic Map