siRNA Details
| Gene Symbol: 1700129O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018999.1 | - | 1 | 363 | ||
| BM942913.1 | - | 1 | 238 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700129O19Rik| Gene Symbol: 1700129O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018999.1 | - | 1 | 363 | ||
| BM942913.1 | - | 1 | 238 | |||