siRNA Details
| Gene Symbol: 1700129O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018999.1 | - | 1 | 482 | ||
| BM942913.1 | - | 1 | 357 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU945787.1 | - | 1 | 497 | Chr. 13 - 49340848 - 49341392 [-] on Build GRCm38 | |

1700129O19Rik
Genomic Map