siRNA Details
| Gene Symbol: 2810429I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034854.1 | - | 2 | 2121 | ||
| AK084238.1 | - | 2 | 2149 | |||
| AK087718.1 | - | 3 | 552 | |||
| Gene Symbol: 4732463B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132490.1 | - | 3 | 2051 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU147524.1 | - | 4 | 583 | Chr. 13 - 3478277 - 3485737 [+] on Build GRCm38 | |

2810429I04Rik
Genomic Map