siRNA Details
| Gene Symbol: Gm3740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158189.1 | - | 2 | 534 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY752961.1 | - | 2 | 534 | Chr. 3 - 85556883 - 85573945 [-] on Build GRCm38 | |

Gm3740
Genomic Map