siRNA Details
| Gene Symbol: Gm40893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034421.1 | - | 1 | 51 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB626463.1 | - | 1 | 49 | Chr. 12 - 98523846 - 98525708 [+] on Build GRCm38 | |

Gm40893
Genomic Map