siRNA Details
| Gene Symbol: 4930594O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016393.1 | - | 2 | 170 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN843869.1 | - | 2 | 154 | Chr. 3 - 37609889 - 37614813 [+] on Build GRCm38 | |

4930594O21Rik
Genomic Map