siRNA Details
| Gene Symbol: 4930412F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015119.1 | - | 3 | 339 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714665.1 | - | 3 | 334 | Chr. 6 - 30896903 - 30943348 [+] on Build GRCm38 | |

4930412F09Rik
Genomic Map