siRNA Details
| Gene Symbol: Gm20036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052505.1 | - | 1 | 1007 | ||
| AK153660.1 | - | 1 | 987 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20036| Gene Symbol: Gm20036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052505.1 | - | 1 | 1007 | ||
| AK153660.1 | - | 1 | 987 | |||