siRNA Details
| Gene Symbol: 4930408O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015114.1 | - | 2 | 370 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714660.1 | - | 2 | 370 | Chr. 12 - 104865330 - 104871525 [+] on Build GRCm38 | |

4930408O17Rik
Genomic Map