siRNA Details
| Gene Symbol: 4930412B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040631.1 | 1 | 42 | |||
| GenBank mRNA | AK015116.1 | - | 1 | 42 | ||
| AK029598.1 | - | 1 | 42 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930412B13Rik| Gene Symbol: 4930412B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040631.1 | 1 | 42 | |||
| GenBank mRNA | AK015116.1 | - | 1 | 42 | ||
| AK029598.1 | - | 1 | 42 | |||