siRNA Details
| Gene Symbol: 4930412B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040631.1 | 3 | 985 | |||
| GenBank mRNA | AK015116.1 | - | 3 | 985 | ||
| AK029598.1 | - | 3 | 985 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV260150.2 | - | 3 | 701 | Chr. 2 - 117822212 - 117909134 [+] on Build GRCm38 | |

4930412B13Rik
Genomic Map