siRNA Details
| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK149226.1 | - | 2 | 234 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742096.1 | - | 2 | 234 | Chr. 8 - 9430007 - 9770104 [-] on Build GRCm38 | |

Fam155a
Genomic Map